5-Methoxyindol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methoxy-1H-indole, Indol-5-yl methyl ether
OR10184-25G
OR10184-5G
OR10184-100G
APOSOR10184-25GEA
43.5
EUR
APOSOR10184-25G
APOSOR10184-5G
APOSOR10184-100G
5-Methoxyindol
5-Methoxyindol
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Siedepunkt: 176…178 °C/17mmHg Schmelzpunkt: 52…58 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005674 CAS-Nummer: 1006-94-6 EINECS: 213-745-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...