Methyl-5-indolcarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 1H-indole-5-carboxylate
OR10190-25G
APOSOR10190-25GEA
0
EUR
APOSOR10190-25G
Methyl-5-indolcarboxylat
Methyl-5-indolcarboxylat
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Schmelzpunkt: 126…128 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00153023 CAS-Nummer: 1011-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...