5-Chlorindol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-1H-indole

OR10199-25G OR10199-5G OR10199-10G
APOSOR10199-25GEA 78.3 EUR
APOSOR10199-25G APOSOR10199-5G APOSOR10199-10G
5-Chlorindol 97%
5-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151,6 g/mol
Schmelzpunkt: 69…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005672
CAS-Nummer: 17422-32-1
EINECS: 241-448-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR