Imidazo[1,2-c]pyrimidin-5(1H)-one 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR1022-10G
OR1022-1G
OR1022-5G
APOSOR1022-10GEA
1250
EUR
APOSOR1022-10G
APOSOR1022-1G
APOSOR1022-5G
Imidazo[1,2-c]pyrimidin-5(1H)-one 95+%
Imidazo[1,2-c]pyrimidin-5(1H)-one
Formel:
C₆H₅N₃O Schmelzpunkt: 280…281 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06800801 CAS-Nummer: 849035-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...