N-Methyltetrahydro-2H-pyran-4-amine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methylamino)tetrahydro-2H-pyran

OR1026-1G OR1026-5G OR1026-10G
APOSOR1026-1GEA 40.6 EUR
APOSOR1026-1G APOSOR1026-5G APOSOR1026-10G
N-Methyltetrahydro-2H-pyran-4-amine 95%
N-Methyltetrahydro-2H-pyran-4-amine
Formel: C₆H₁₃NO
Molecular Weight: 115.1700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06658492
CAS-Nummer: 220641-87-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR