1,5-Naphthyridine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR1027-1G OR1027-5G
APOSOR1027-1GEA 473 EUR
APOSOR1027-1G APOSOR1027-5G
1,5-Naphthyridine-2-carboxylic acid 97%
1,5-Naphthyridine-2-carboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06801138
CAS-Nummer: 49850-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR