5-Nitro-2-benzofurancarbonsäure

Lieferant: Apollo Scientific
Achtung

OR102840-1G OR102840-25G OR102840-5G
APOSOR102840-1GEA 24.8 EUR
APOSOR102840-1G APOSOR102840-25G APOSOR102840-5G
5-Nitro-2-benzofurancarbonsäure
5-Nitrobenzofuran-2-carboxylic acid
Formel: C₉H₅NO₅
Molecular Weight: 207,1397 g/mol
Schmelzpunkt: 279…284 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060513
CAS-Nummer: 10242-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR