2-(1H-Pyrazol-4-yl)pyridin 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR1029-1G
OR1029-5G
APOSOR1029-1GEA
108
EUR
APOSOR1029-1G
APOSOR1029-5G
2-(1H-Pyrazol-4-yl)pyridin 95%
2-(1H-Pyrazol-4-yl)pyridin
Formel:
C₈H₇N₃ Molecular Weight: 145,1613 g/mol Schmelzpunkt: 134…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02579318 CAS-Nummer: 439106-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...