2,2,2-Trichloroethyl acrylate

Lieferant: Apollo Scientific

Synonyme: Acrylic acid, 2,2,2-trichloroethyl ester

OR10304-1G OR10304-25G OR10304-5G
APOSOR10304-1GEA 94.4 EUR
APOSOR10304-1G APOSOR10304-25G APOSOR10304-5G
2,2,2-Trichloroethyl acrylate
2,2,2-Trichloroethyl acrylate
Formel: C₅H₅Cl₃O₂
Molecular Weight: 203,45 g/mol
Siedepunkt: 31,5 °C/0.5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04038361
CAS-Nummer: 44925-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR