4-Acetylphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

OR10313-25G OR10313-5G OR10313-100G
APOSOR10313-25GEA 43.5 EUR
APOSOR10313-25G APOSOR10313-5G APOSOR10313-100G
4-Acetylphenylboronsäure 98%
4-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163,97 g/mol
Schmelzpunkt: 243…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01074667
CAS-Nummer: 149104-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR