4-Carbamoylphenylboronic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-carbamoylphenyl)boronic Acid, 4-Aminocarbonylphenylboronic acid
OR10325-25G
OR10325-5G
OR10325-1G
APOSOR10325-25GEA
82.8
EUR
APOSOR10325-25G
APOSOR10325-5G
APOSOR10325-1G
4-Carbamoylphenylboronic acid 96%
4-Carbamoylphenylboronic acid
Formel:
C₇H₈BNO₃ Molecular Weight: 164,955 g/mol Schmelzpunkt: 229…234 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411940 CAS-Nummer: 123088-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...