(3-Carbamoylphenyl)boronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-carbamoylphenyl)boronic acid, 3-Aminocarbonylphenylboronic acid, 3-carbamoylbenzeneboronic acid, 3-Carbamoylphenylboronic acid, 3-Carbamoylbenzeneboronic acid

OR10326-25G OR10326-5G OR10326-100G
APOSOR10326-25GEA 62.5 EUR
APOSOR10326-25G APOSOR10326-5G APOSOR10326-100G
(3-Carbamoylphenyl)boronic acid 98%
(3-Carbamoylphenyl)boronic acid
Formel: C₇H₈BNO₃
Molecular Weight: 164,955 g/mol
Schmelzpunkt: 232…238 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411948
CAS-Nummer: 351422-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR