5-Amino-1-methyl-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR103267-1G OR103267-5G OR103267-25G
APOSOR103267-1GEA 58 EUR
APOSOR103267-1G APOSOR103267-5G APOSOR103267-25G
5-Amino-1-methyl-1H-pyrazole-4-carboxylic acid
5-Amino-1-methyl-1H-pyrazole-4-carboxylic acid
Formel: C₅H₇N₃O₂
Molecular Weight: 141.1280 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00973833
CAS-Nummer: 4058-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR