5-Aminooxindol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Aminoindolin-2-one, 5-Amino-1,3-dihydro-indol-2-one, 5-Amino-2-indolinone, 5-Aminooxindole

OR10335-5G
APOSOR10335-5GEA 127 EUR
APOSOR10335-5G
5-Aminooxindol
5-Aminooxindol
Formel: C₈H₈N₂O
Molecular Weight: 148,16 g/mol
Schmelzpunkt: 182…186 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179603
CAS-Nummer: 20876-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR