4-(Benzylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(Benzylcarbamoyl)phenyl)boronic acid, 4-(Benzylcarbamoyl)benzeneboronic acid, 4-(N-Benzylaminocarbonyl)phenylboronic acid
OR10355-1G
OR10355-5G
APOSOR10355-1GEA
26.1
EUR
APOSOR10355-1G
APOSOR10355-5G
4-(Benzylcarbamoyl)phenylboronsäure 98%
4-(Benzylcarbamoyl)phenylboronsäure
Formel:
C₁₄H₁₄BNO₃ Molecular Weight: 255,08 g/mol Schmelzpunkt: 212…218 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411953 CAS-Nummer: 252663-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...