3-(Benzyloxycarbonyl)phenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carbobenzoxybenzeneboronic acid, (3-((Benzyloxy)carbonyl)phenyl)boronic acid, 3-Carbobenzoxyphenylboronic acid, 3-(Benzyloxycarbonyl)benzeneboronic acid

OR10358-5G
APOSOR10358-5GEA 128 EUR
APOSOR10358-5G
3-(Benzyloxycarbonyl)phenylboronic acid 98%
3-(Benzyloxycarbonyl)phenylboronic acid
Formel: C₁₄H₁₃BO₄
Molecular Weight: 256,06 g/mol
Schmelzpunkt: 138…146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179445
CAS-Nummer: 380430-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR