4-(Benzyloxycarbonyl)benzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-((Benzyloxy)carbonyl)phenyl)boronic acid, 4-Benzyloxycarbonylphenylboronic acid, 4-Carbobenzoxybenzeneboronic acid, 4-(Benzyloxycarbonyl)benzeneboronic acid, 4-Carbobenzoxyphenylboronic acid, 4-(Benzyloxycarbonyl)phenylboronic acid

OR10359-1G OR10359-25G OR10359-5G
APOSOR10359-1GEA 37.7 EUR
APOSOR10359-1G APOSOR10359-25G APOSOR10359-5G
4-(Benzyloxycarbonyl)benzeneboronic acid 97%
4-(Benzyloxycarbonyl)benzeneboronic acid
Formel: C₁₄H₁₃BO₄
Molecular Weight: 256.06 g/mol
Schmelzpunkt: 187…192 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179442
CAS-Nummer: 184000-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR