4-(2-Aminopyrimidin-4-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR103635-1G OR103635-250MG
APOSOR103635-1GEA 406 EUR
APOSOR103635-1G APOSOR103635-250MG
4-(2-Aminopyrimidin-4-yl)benzoic acid
4-(2-Aminopyrimidin-4-yl)benzoic acid
Formel: C₁₁H₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03407445
CAS-Nummer: 216959-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR