4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR103636-10G OR103636-1G OR103636-5G
APOSOR103636-10GEA 455 EUR
APOSOR103636-10G APOSOR103636-1G APOSOR103636-5G
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
Formel: C₉H₁₀O₂S
Molecular Weight: 182,24 g/mol
Schmelzpunkt: 192…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00464469
CAS-Nummer: 40133-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR