2-Brom-5-formylthiazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromothiazole-5-carboxaldehyde
OR10364-5G
OR10364-1G
APOSOR10364-5GEA
116
EUR
APOSOR10364-5G
APOSOR10364-1G
2-Brom-5-formylthiazol 98%
2-Brom-5-formylthiazol
Formel:
C₄H₂BrNOS Molecular Weight: 192.03 g/mol Schmelzpunkt: 92…96 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03788567 CAS-Nummer: 464192-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...