7-Nitro-2,3-dihydro-1-benzofuran-5-sulphonoylchloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,3-Dihydro-7-nitrobenzo[b]furan-5-sulphonyl chloride
OR103649-500MG
OR103649-1G
APOSOR103649-500MGEA
132
EUR
APOSOR103649-500MG
APOSOR103649-1G
7-Nitro-2,3-dihydro-1-benzofuran-5-sulphonoylchloride
7-Nitro-2,3-dihydro-1-benzofuran-5-sulphonoylchloride
Formel:
C₈H₆ClNO₅S Molecular Weight: 263,66 g/mol Schmelzpunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03408011 CAS-Nummer: 874781-18-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...