2-Bromimidazol 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Brom-1H-imidazol

OR10366-10G OR10366-1G OR10366-5G
APOSOR10366-10GEA 145 EUR
APOSOR10366-10G APOSOR10366-1G APOSOR10366-5G
2-Bromimidazol 98%
2-Bromoimidazole
Formel: C₃H₃BrN₂
Molecular Weight: 146,97 g/mol
Schmelzpunkt: 197…200 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179526
CAS-Nummer: 16681-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR