4-Brom-2-methylimidazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Brom-2-methyl-1H-imidazol
OR10370-25G
OR10370-5G
APOSOR10370-25GEA
241
EUR
APOSOR10370-25G
APOSOR10370-5G
4-Brom-2-methylimidazol 98%
4-Bromo-2-methylimidazole
Formel:
C₄H₅BrN₂ Molecular Weight: 161 g/mol Schmelzpunkt: 158…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411959 CAS-Nummer: 16265-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...