6-Bromoxindol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

OR10384-1G OR10384-25G OR10384-5G
APOSOR10384-1GEA 21.9 EUR
APOSOR10384-1G APOSOR10384-25G APOSOR10384-5G
6-Bromoxindol 97%
6-Bromoxindol
Formel: C₈H₆BrNO
Molecular Weight: 212.05 g/mol
Schmelzpunkt: 217…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179605
CAS-Nummer: 99365-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR