7-Bromoxindol
Lieferant: Apollo Scientific
|
Achtung
|
OR10385-1G
OR10385-250MG
OR10385-5G
APOSOR10385-1GEA
0
EUR
APOSOR10385-1G
APOSOR10385-250MG
APOSOR10385-5G
7-Bromoxindol
7-Bromoxindol
Formel:
C₈H₆BrNO Molecular Weight: 212,0433 g/mol Schmelzpunkt: 209 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179613 CAS-Nummer: 320734-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...