3-(tert-Butoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(tert-Butoxycarbonyl)benzeneboronic acid, 3-tert-Butoxycarbonylphenylboronic acid

OR10391-5G OR10391-100G OR10391-25G
APOSOR10391-5GEA 21.9 EUR
APOSOR10391-5G APOSOR10391-100G APOSOR10391-25G
3-(tert-Butoxycarbonyl)phenylboronsäure 98%
3-(tert-Butoxycarbonyl)phenylboronsäure
Formel: C₁₁H₁₅BO₄
Molecular Weight: 222.05 g/mol
Schmelzpunkt: 96…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01630855
CAS-Nummer: 220210-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR