Oxindol-5-carbonsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Oxoindoline-5-carboxylic Acid, 2-Oxindole-5-carboxylic acid

OR10403-1G OR10403-250MG OR10403-5G
APOSOR10403-1GEA 59.6 EUR
APOSOR10403-1G APOSOR10403-250MG APOSOR10403-5G
Oxindol-5-carbonsäure 97%
Oxindol-5-carbonsäure
Formel: C₉H₇NO₃
Molecular Weight: 177,1600 g/mol
Schmelzpunkt: 310…316 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411961
CAS-Nummer: 102359-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR