2-Carboxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Boronobenzoic acid, 2-Carboxybenzeneboronic acid
OR10404-25G
OR10404-5G
OR10404-10G
APOSOR10404-25GEA
267
EUR
APOSOR10404-25G
APOSOR10404-5G
APOSOR10404-10G
2-Carboxyphenylboronsäure
2-Carboxyphenylboronsäure
Formel:
C₇H₇BO₄ Molecular Weight: 165,94 g/mol Schmelzpunkt: 220 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01318118 CAS-Nummer: 149105-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...