4-Carboxyphenylboronsäurepinakolester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
OR10406-25G
OR10406-5G
OR10406-100G
APOSOR10406-25GEA
33.5
EUR
APOSOR10406-25G
APOSOR10406-5G
APOSOR10406-100G
4-Carboxyphenylboronsäurepinakolester 98%
4-Carboxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ Molecular Weight: 248,08 g/mol Schmelzpunkt: 227…231 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01863710 CAS-Nummer: 180516-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...