2-Chlor-4-methoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-4-methoxybenzeneboronic acid
OR10412-5G
OR10412-1G
OR10412-25G
APOSOR10412-5GEA
37.7
EUR
APOSOR10412-5G
APOSOR10412-1G
APOSOR10412-25G
2-Chlor-4-methoxyphenylboronsäure
2-Chlor-4-methoxyphenylboronsäure
Formel:
C₇H₈BClO₃ Molecular Weight: 186,40 g/mol Schmelzpunkt: 202 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411935 CAS-Nummer: 219735-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...