2-Chlor-4-methylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-methylbenzeneboronic acid

OR10414-1G OR10414-25G OR10414-5G
APOSOR10414-1GEA 21.9 EUR
APOSOR10414-1G APOSOR10414-25G APOSOR10414-5G
2-Chlor-4-methylphenylboronsäure
2-Chlor-4-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170,40 g/mol
Schmelzpunkt: 198…202 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411936
CAS-Nummer: 145349-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR