4-(Cyanomethyl)phenylboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Cyanomethyl)benzeneboronic acid

OR10422-1G OR10422-5G OR10422-10G
APOSOR10422-1GEA 40.6 EUR
APOSOR10422-1G APOSOR10422-5G APOSOR10422-10G
4-(Cyanomethyl)phenylboronic acid 97%
4-(Cyanomethyl)phenylboronic acid
Formel: C₈H₈BNO₂
Schmelzpunkt: 196…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632200
CAS-Nummer: 91983-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR