3-(Cyanomethyl)benzeneboronic acid pinacol ester 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetonitrile, 2-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile

OR10423-25G OR10423-5G
APOSOR10423-25GEA 275 EUR
APOSOR10423-25G APOSOR10423-5G
3-(Cyanomethyl)benzeneboronic acid pinacol ester 96%
3-(Cyanomethyl)benzeneboronic acid pinacol ester
Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243,113 g/mol
Schmelzpunkt: 44…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179458
CAS-Nummer: 396131-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR