3-(Cyanomethyl)benzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(Cyanomethyl)phenyl)boronic acid

OR10425-1G OR10425-250MG OR10425-5G
APOSOR10425-1GEA 261 EUR
APOSOR10425-1G APOSOR10425-250MG APOSOR10425-5G
3-(Cyanomethyl)benzeneboronic acid 97%
3-(Cyanomethyl)benzeneboronic acid
Formel: C₈H₈BNO₂
Molecular Weight: 160.97 g/mol
Schmelzpunkt: 85…88 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03788026
CAS-Nummer: 220616-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR