2-Cyanphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Dihydroxy(2-cyanphenyl)boran, 2-Cyanbenzolboronsäure, (2-Cyanphenyl)metaborsäure

OR10429-5G OR10429-10G OR10429-1G
APOSOR10429-5GEA 49.3 EUR
APOSOR10429-5G APOSOR10429-10G APOSOR10429-1G
2-Cyanphenylboronsäure 98%
2-Cyanphenylboronsäure

Employed in a rhodium-catalyzed [3+2] annulation with alkynes leading to substituted indenones .

Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
Schmelzpunkt: 250…254 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD01632208
CAS-Nummer: 138642-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR