4-(Cyclohexylcarbamoyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Cyclohexylaminocarbonyl)phenylboronic acid

OR10430-1G OR10430-5G OR10430-25G
APOSOR10430-1GEA 52.2 EUR
APOSOR10430-1G APOSOR10430-5G APOSOR10430-25G
4-(Cyclohexylcarbamoyl)phenylboronsäure
4-(Cyclohexylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₈BNO₃
Schmelzpunkt: 174…178 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411956
CAS-Nummer: 762262-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR