4-(Cyclopropylcarbamoyl)phenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Cyclopropylcarbamoyl)benzeneboronic acid, 4-(Cyclopropylaminocarbonyl)phenylboronic acid
OR10432-5G
OR10432-25G
APOSOR10432-5GEA
165
EUR
APOSOR10432-5G
APOSOR10432-25G
4-(Cyclopropylcarbamoyl)phenylboronsäure 97%
4-(Cyclopropylcarbamoyl)phenylboronsäure
Formel:
C₁₀H₁₂BNO₃ Molecular Weight: 205,02 g/mol Schmelzpunkt: 202…206 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03788419 CAS-Nummer: 515140-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...