3,4-Dichlorphenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR10445-100G OR10445-25G OR10445-250G
APOSOR10445-100GEA 108 EUR
APOSOR10445-100G APOSOR10445-25G APOSOR10445-250G
3,4-Dichlorphenylboronsäure
3,4-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190,82 g/mol
Schmelzpunkt: 280…285 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01074646
CAS-Nummer: 151169-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR