3-(Dimethylcarbamoyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(N,N-Dimethylaminocarbonyl)phenylboronic acid, 3-(Dimethylcarbamoyl)benzeneboronic acid, (3-(Dimethylcarbamoyl)phenyl)boronic acid
OR10458-1G
OR10458-5G
APOSOR10458-1GEA
0
EUR
APOSOR10458-1G
APOSOR10458-5G
3-(Dimethylcarbamoyl)phenylboronsäure
3-(Dimethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₂BNO₃ Molecular Weight: 193.01 g/mol Schmelzpunkt: 122…128 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03412066 CAS-Nummer: 373384-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...