2,6-Dimethylphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(2,6-dimethylphenyl)boran, (2,6-Dimethylphenyl)metaborsäure, 2,6-Dimethylbenzolboronsäure
OR10461-1G
OR10461-25G
OR10461-5G
APOSOR10461-1GEA
21.9
EUR
APOSOR10461-1G
APOSOR10461-25G
APOSOR10461-5G
2,6-Dimethylphenylboronsäure 97%
2,6-Dimethylphenylboronsäure
Formel:
C₈H₁₁BO₂ Molecular Weight: 149,98 g/mol Schmelzpunkt: 120…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01009693 CAS-Nummer: 100379-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...