2,5-Dimethylphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR10463-25G
OR10463-5G
APOSOR10463-25GEA
33.5
EUR
APOSOR10463-25G
APOSOR10463-5G
2,5-Dimethylphenylboronsäure 98%
2,5-Dimethylphenylboronsäure
Formel:
C₈H₁₁BO₂ Molecular Weight: 149,98 g/mol Schmelzpunkt: 186…191 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01863525 CAS-Nummer: 85199-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...