1-Ethoxymethyl-2-iodimidazol

Lieferant: Apollo Scientific
Achtung

OR1047-1G OR1047-250MG
APOSOR1047-1GEA 102 EUR
APOSOR1047-1G APOSOR1047-250MG
1-Ethoxymethyl-2-iodimidazol
1-Ethoxymethyl-2-iodimidazol
Formel: C₆H₉IN₂O
Molecular Weight: 252.05 g/mol
Schmelzpunkt: 51…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788568
CAS-Nummer: 146697-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR