4-Ethylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Ethylphenyl)metaborsäure, 4-Ethylbenzolboronsäure, Dihydroxy(4-ethylphenyl)boran

OR10478-100G OR10478-25G
APOSOR10478-100GEA 58 EUR
APOSOR10478-100G APOSOR10478-25G
4-Ethylphenylboronsäure
4-Ethylphenylboronsäure
Formel: C₈H₁₁BO₂
Molecular Weight: 149.98 g/mol
Schmelzpunkt: 150…155 °C
Dichte: 1.125
Lagertemperatur: Kühlschrank
MDL: MFCD00859377
CAS-Nummer: 63139-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR