2-Formyl-3-thiophenboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Formylthiophene-3-boronic acid, (2-Formylthiophen-3-yl)boronic acid

OR10484-25G OR10484-5G
APOSOR10484-25GEA 516 EUR
APOSOR10484-25G APOSOR10484-5G
2-Formyl-3-thiophenboronsäure 97%
2-Formyl-3-thiophenboronsäure
Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01075678
CAS-Nummer: 4347-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR