5-Formyl-2-thiophenboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid
OR10486-100G
OR10486-25G
OR10486-5G
APOSOR10486-100GEA
215
EUR
APOSOR10486-100G
APOSOR10486-25G
APOSOR10486-5G
5-Formyl-2-thiophenboronsäure
5-Formyl-2-thiophenboronsäure
Formel:
C₅H₅BO₃S Molecular Weight: 155,97 g/mol Schmelzpunkt: 132…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093666 CAS-Nummer: 4347-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...