5-Indolcarbaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Indol-5-carboxaldehyd
OR10488-25G
OR10488-5G
APOSOR10488-25GEA
198
EUR
APOSOR10488-25G
APOSOR10488-5G
5-Indolcarbaldehyd 98%
5-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 96…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093664 CAS-Nummer: 1196-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...