6-Indolcarbaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Indol-6-carboxaldehyd
OR10489-25G
APOSOR10489-25GEA
207
EUR
APOSOR10489-25G
6-Indolcarbaldehyd
6-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 127…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179596 CAS-Nummer: 1196-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...