4,6-Dimethoxysalicylaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-4,6-dimethoxybenzaldehyde, 4,6-Dimethoxy-2-hydroxybenzaldehyde

OR10496-100G OR10496-25G
APOSOR10496-100GEA 382 EUR
APOSOR10496-100G APOSOR10496-25G
4,6-Dimethoxysalicylaldehyd 98%
4,6-Dimethoxysalicylaldehyd
Formel: C₉H₁₀O₄
Molecular Weight: 182.1733 g/mol
Schmelzpunkt: 60…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003328
CAS-Nummer: 708-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR