4-Brom-3-picolin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-3-methylpyridine hydrochloride

OR1050-1G OR1050-250MG OR1050-5G
APOSOR1050-1GEA 24.8 EUR
APOSOR1050-1G APOSOR1050-250MG APOSOR1050-5G
4-Brom-3-picolin Hydrochlorid
4-Brom-3-picolin Hydrochlorid
Formel: C₆H₇BrClN
Molecular Weight: 208.483 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04966778
CAS-Nummer: 40899-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR