4-Iod-1-methylimidazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Iod-1-methyl-1H-imidazol
OR10511-1G
OR10511-5G
OR10511-10G
APOSOR10511-1GEA
24.8
EUR
APOSOR10511-1G
APOSOR10511-5G
APOSOR10511-10G
4-Iod-1-methylimidazol 97%
4-Iodo-1-methylimidazole
Formel:
C₄H₅IN₂ Molecular Weight: 208 g/mol Schmelzpunkt: 51…54 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01632214 CAS-Nummer: 71759-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...